Assay Details
| Gene Symbol: limk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042691.2 | NP_001036156.1 | 5 - 6 | 632 | 79 | |
| XM_005169645.4 | XP_005169702.1 | 3 - 4 | 538 | 79 | ||
| XM_005169646.4 | XP_005169703.1 | 4 - 5 | 577 | 79 | ||
| GenBank mRNA | BC163263.1 | - | 5 - 6 | 632 | 79 | |
| DQ679229.1 | - | 5 - 6 | 632 | 79 | ||
More Information
Set Membership: |
|

limk1a
Genomic Map