Assay Details
Gene Symbol: nptxrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044314.1 | NP_001037779.1 | 1 - 2 | 822 | 88 | |
GenBank mRNA | BC067657.1 | - | 1 - 2 | 696 | 88 | |
BC109464.1 | - | 2 - 3 | 734 | 88 | ||
BC131861.1 | - | 1 - 2 | 822 | 88 | ||
BC171437.1 | - | 1 - 2 | 811 | 88 | ||
BC171439.1 | - | 1 - 2 | 811 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
