Assay Details
Gene Symbol: oxr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045203.1 | NP_001038668.1 | 12 - 13 | 2112 | 75 | |
XM_005159458.3 | XP_005159515.1 | 15 - 16 | 2506 | 75 | ||
XM_005159459.3 | XP_005159516.1 | 14 - 15 | 2425 | 75 | ||
XM_009294057.3 | XP_009292332.1 | 5 - 6 | 917 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
