Assay Details
Gene Symbol: pdpk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077344.1 | NP_001070812.1 | 6 - 7 | 828 | 75 | |
GenBank mRNA | BC091466.1 | - | 7 - 8 | 770 | 75 | |
BC124736.1 | - | 6 - 7 | 828 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
