Assay Details
| Gene Symbol: trim3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080168.1 | NP_001073637.1 | 2 - 3 | 345 | 151 | |
| XM_005155377.4 | XP_005155434.1 | 2 - 3 | 339 | 151 | ||
| XM_005155378.4 | XP_005155435.1 | 2 - 3 | 365 | 151 | ||
| XM_009305315.3 | XP_009303590.1 | 3 - 4 | 391 | 151 | ||
| XM_021479176.1 | XP_021334851.1 | 2 - 3 | 321 | 151 | ||
| GenBank mRNA | BC129039.1 | - | 2 - 3 | 345 | 151 | |
More Information
Set Membership: |
|

trim3b
Genomic Map