Assay Details
| Gene Symbol: smarcc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089367.1 | NP_001082836.1 | 16 - 17 | 1515 | 72 | |
| GenBank mRNA | BC134882.1 | - | 16 - 17 | 1515 | 72 | |
More Information
Set Membership: |
|

smarcc1b
Genomic Map