Assay Details
| Gene Symbol: smarcc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089367.1 | NP_001082836.1 | 26 - 27 | 2830 | 71 | |
| GenBank mRNA | BC134882.1 | - | 26 - 27 | 2830 | 71 | |
More Information
Set Membership: |
|

smarcc1b
Genomic Map