Assay Details
| Gene Symbol: cyp24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089458.1 | NP_001082927.1 | 10 - 11 | 1448 | 75 | |
| GenBank mRNA | BC133740.1 | - | 10 - 11 | 1448 | 75 | |
More Information
Set Membership: |
|

cyp24a1
Genomic Map