Assay Details
| Gene Symbol: ttll6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105524.1 | NP_001098994.1 | 2 - 3 | 224 | 75 | |
| GenBank mRNA | EU124004.1 | - | 2 - 3 | 224 | 75 | |
More Information
Set Membership: |
|

ttll6
Genomic Map