Assay Details
Gene Symbol: parvab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110471.1 | NP_001103941.1 | 12 - 13 | 1106 | 76 | |
GenBank mRNA | BC134924.1 | - | 12 - 13 | 1106 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
