Assay Details
Gene Symbol: parvab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110471.1 | NP_001103941.1 | 1 - 2 | 202 | 80 | |
GenBank mRNA | BC134924.1 | - | 1 - 2 | 202 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
