Assay Details
| Gene Symbol: cox19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110540.2 | NP_001104010.1 | 1 - 2 | 194 | 102 | |
| GenBank mRNA | BC139534.1 | - | 1 - 2 | 155 | 102 | |
| BC171515.1 | - | 1 - 2 | 165 | 102 | ||
| BC171517.1 | - | 1 - 2 | 165 | 102 | ||
| EH445842.1 | - | 1 - 2 | 226 | 102 | ||
More Information
Set Membership: |
|

cox19
Genomic Map