Assay Details
Gene Symbol: lmnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152972.2 | NP_694504.2 | 6 - 7 | 1082 | 68 | |
XM_005155556.4 | XP_005155613.1 | 6 - 7 | 1031 | 68 | ||
GenBank mRNA | AJ250201.1 | - | 5 - 6 | 942 | 68 | |
BC044402.1 | - | 6 - 7 | 1082 | 68 | ||
BC068414.1 | - | 5 - 6 | 939 | 68 | ||
BC165769.1 | - | 5 - 6 | 976 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
