Assay Details
| Gene Symbol: myo7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152983.1 | NP_694515.1 | 19 - 20 | 2447 | 127 | |
| XM_021467573.1 | XP_021323248.1 | 20 - 21 | 2598 | 127 | ||
| GenBank mRNA | AJ404001.1 | - | 19 - 20 | 2447 | 127 | |
| AJ404002.1 | - | 18 - 19 | 2327 | 127 | ||
| BC163570.1 | - | 18 - 19 | 2335 | 127 | ||
More Information
Set Membership: |
|

myo7aa
Genomic Map