Assay Details
Gene Symbol: smad3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175083.2 | NP_778258.1 | 2 - 3 | 411 | 118 | |
GenBank mRNA | AY134490.1 | - | 2 - 3 | 411 | 118 | |
BC081628.1 | - | 2 - 3 | 549 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
