Assay Details
Gene Symbol: smad3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175083.2 | NP_778258.1 | 5 - 6 | 660 | 74 | |
GenBank mRNA | AY134490.1 | - | 5 - 6 | 660 | 74 | |
BC081628.1 | - | 5 - 6 | 798 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
