Assay Details
| Gene Symbol: adra2da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194364.2 | NP_919345.2 | 1 | 1155 | 139 | |
| GenBank mRNA | BC129157.1 | - | 1 | 1155 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

adra2da| Gene Symbol: adra2da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194364.2 | NP_919345.2 | 1 | 1155 | 139 | |
| GenBank mRNA | BC129157.1 | - | 1 | 1155 | 139 | |
Set Membership: |
|