Assay Details
| Gene Symbol: p2rx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198985.1 | NP_945336.1 | 7 - 8 | 747 | 92 | |
| GenBank mRNA | AY292648.1 | - | 7 - 8 | 747 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

p2rx8| Gene Symbol: p2rx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198985.1 | NP_945336.1 | 7 - 8 | 747 | 92 | |
| GenBank mRNA | AY292648.1 | - | 7 - 8 | 747 | 92 | |
Set Membership: |
|