Assay Details
| Gene Symbol: pdap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200210.1 | NP_956504.1 | 3 - 4 | 478 | 88 | |
| GenBank mRNA | BC045997.1 | - | 3 - 4 | 478 | 88 | |
More Information
Set Membership: |
|

pdap1b
Genomic Map