Assay Details
| Gene Symbol: cxxc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200599.1 | NP_956893.1 | 10 - 11 | 1431 | 62 | |
| GenBank mRNA | BC056775.1 | - | 10 - 11 | 1431 | 62 | |
More Information
Set Membership: |
|

cxxc1a
Genomic Map