Assay Details
Gene Symbol: abi1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328427.1 | NP_001315356.1 | 5 - 6 | 818 | 85 | |
XM_005162489.4 | XP_005162546.1 | 6 - 7 | 834 | 85 | ||
XM_005162496.4 | XP_005162553.1 | 5 - 6 | 819 | 85 | ||
XM_005162498.4 | XP_005162555.1 | 6 - 7 | 833 | 85 | ||
XM_005162499.4 | XP_005162556.1 | 5 - 6 | 818 | 85 | ||
XM_009297227.3 | XP_009295502.1 | 6 - 7 | 835 | 85 | ||
XM_009297228.3 | XP_009295503.1 | 5 - 6 | 819 | 85 | ||
XM_009297229.3 | XP_009295504.1 | 6 - 7 | 834 | 85 | ||
GenBank mRNA | BC059524.1 | - | 5 - 6 | 555 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
