Assay Details
Gene Symbol: abi1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328427.1 | NP_001315356.1 | 6 - 7 | 917 | 75 | |
XM_005162489.4 | XP_005162546.1 | 7 - 8 | 933 | 75 | ||
GenBank mRNA | BC059524.1 | - | 6 - 7 | 654 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
