Assay Details
| Gene Symbol: adrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201013.3 | NP_957307.3 | 3 - 4 | 476 | 83 | |
| XM_005161962.4 | XP_005162019.1 | 3 - 4 | 486 | 83 | ||
| XM_021469854.1 | XP_021325529.1 | 3 - 4 | 486 | 83 | ||
| GenBank mRNA | BC045915.1 | - | 3 - 4 | 433 | 83 | |
| BC066391.1 | - | 3 - 4 | 452 | 83 | ||
| BC154825.1 | - | 3 - 4 | 412 | 83 | ||
| EH594482.1 | - | 3 - 4 | 503 | 83 | ||
More Information
Set Membership: |
|

adrm1
Genomic Map