Assay Details
| Gene Symbol: etf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201460.2 | NP_958868.1 | 7 - 8 | 1051 | 74 | |
| XM_021479101.1 | XP_021334776.1 | 7 - 8 | 1066 | 74 | ||
| GenBank mRNA | AY398343.1 | - | 7 - 8 | 1051 | 74 | |
| BC044515.1 | - | 7 - 8 | 1045 | 74 | ||
| BC066583.1 | - | 7 - 8 | 969 | 74 | ||
More Information
Set Membership: |
|

etf1b
Genomic Map