Assay Details
| Gene Symbol: arcn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201993.1 | NP_973722.1 | 3 - 4 | 554 | 83 | |
| GenBank mRNA | BC050499.1 | - | 3 - 4 | 554 | 83 | |
| BC075749.1 | - | 3 - 4 | 513 | 83 | ||
More Information
Set Membership: |
|

arcn1b
Genomic Map