Assay Details
Gene Symbol: arcn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201993.1 | NP_973722.1 | 5 - 6 | 927 | 79 | |
GenBank mRNA | BC050499.1 | - | 5 - 6 | 927 | 79 | |
BC075749.1 | - | 5 - 6 | 886 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
