Assay Details
| Gene Symbol: arcn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201993.1 | NP_973722.1 | 8 - 9 | 1356 | 87 | |
| GenBank mRNA | BC050499.1 | - | 8 - 9 | 1356 | 87 | |
| BC075749.1 | - | 8 - 9 | 1315 | 87 | ||
More Information
Set Membership: |
|

arcn1b
Genomic Map