Assay Details
| Gene Symbol: glud1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212576.1 | NP_997741.1 | 12 - 13 | 1632 | 75 | |
| GenBank mRNA | BC066370.1 | - | 12 - 13 | 1632 | 75 | |
More Information
Set Membership: |
|

glud1a
Genomic Map