Assay Details
| Gene Symbol: slc20a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212588.1 | NP_997753.1 | 4 - 5 | 705 | 69 | |
| XM_005155625.4 | XP_005155682.1 | 5 - 6 | 1385 | 69 | ||
| GenBank mRNA | BC057497.1 | - | 4 - 5 | 705 | 69 | |
More Information
Set Membership: |
|

slc20a1b
Genomic Map