Assay Details
| Gene Symbol: slc20a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212588.1 | NP_997753.1 | 7 - 8 | 1594 | 65 | |
| XM_005155625.4 | XP_005155682.1 | 8 - 9 | 2274 | 65 | ||
| GenBank mRNA | BC057497.1 | - | 7 - 8 | 1594 | 65 | |
More Information
Set Membership: |
|

slc20a1b
Genomic Map