Assay Details
| Gene Symbol: plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212690.1 | NP_997855.1 | 7 - 8 | 1106 | 54 | |
| GenBank mRNA | BC055539.1 | - | 7 - 8 | 1106 | 54 | |
More Information
Set Membership: |
|

plaa
Genomic Map