Assay Details
| Gene Symbol: ssrp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212802.2 | NP_997967.2 | 5 - 6 | 702 | 72 | |
| GenBank mRNA | BC056311.1 | - | 5 - 6 | 608 | 72 | |
| CT594375.2 | - | 5 - 6 | 663 | 72 | ||
| Gene Symbol: ssrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK238113.1 | - | 3 - 4 | 490 | 72 | |
More Information
Set Membership: |
|

ssrp1a
Genomic Map