Assay Details
| Gene Symbol: tead1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212847.2 | NP_998012.1 | 13 - 14 | 1379 | 102 | |
| XM_005174314.3 | XP_005174371.1 | 12 - 13 | 1494 | 102 | ||
| XM_009297895.3 | XP_009296170.1 | 11 - 12 | 1392 | 102 | ||
| GenBank mRNA | BC055211.1 | - | 13 - 14 | 1380 | 102 | |
| BC097055.1 | - | 12 - 13 | 1444 | 102 | ||
| BC155141.1 | - | 12 - 13 | 1378 | 102 | ||
| BC164522.1 | - | 10 - 11 | 1191 | 102 | ||
More Information
Set Membership: |
|

tead1a
Genomic Map