Assay Details
| Gene Symbol: tead1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212847.2 | NP_998012.1 | 9 - 10 | 782 | 78 | |
| XM_005174314.3 | XP_005174371.1 | 8 - 9 | 897 | 78 | ||
| XM_009297895.3 | XP_009296170.1 | 7 - 8 | 795 | 78 | ||
| GenBank mRNA | BC055211.1 | - | 9 - 10 | 783 | 78 | |
| BC097055.1 | - | 8 - 9 | 847 | 78 | ||
| BC164522.1 | - | 6 - 7 | 594 | 78 | ||
More Information
Set Membership: |
|

tead1a
Genomic Map