Assay Details
| Gene Symbol: tead1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212847.2 | NP_998012.1 | 11 - 12 | 1082 | 85 | |
| XM_005174314.3 | XP_005174371.1 | 10 - 11 | 1197 | 85 | ||
| XM_009297895.3 | XP_009296170.1 | 9 - 10 | 1095 | 85 | ||
| GenBank mRNA | BC055211.1 | - | 11 - 12 | 1083 | 85 | |
| BC155141.1 | - | 10 - 11 | 1081 | 85 | ||
More Information
Set Membership: |
|

tead1a
Genomic Map