Assay Details
| Gene Symbol: amy2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213011.2 | NP_998176.2 | 6 - 7 | 1030 | 74 | |
| GenBank mRNA | BC054669.1 | - | 6 - 7 | 1005 | 74 | |
| BC062867.1 | - | 6 - 7 | 1007 | 74 | ||
More Information
Set Membership: |
|

amy2a
Genomic Map