Assay Details
| Gene Symbol: caprin1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213068.1 | NP_998233.1 | 13 - 14 | 1540 | 72 | |
| XM_021467626.1 | XP_021323301.1 | 13 - 14 | 1557 | 72 | ||
| XM_021467627.1 | XP_021323302.1 | 13 - 14 | 1560 | 72 | ||
| GenBank mRNA | BC047196.1 | - | 13 - 14 | 1540 | 72 | |
More Information
Set Membership: |
|

caprin1b
Genomic Map