Assay Details
| Gene Symbol: bzw1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213092.1 | NP_998257.1 | 6 - 7 | 600 | 88 | |
| XM_005167390.4 | XP_005167447.1 | 6 - 7 | 690 | 88 | ||
| GenBank mRNA | BC044401.1 | - | 6 - 7 | 620 | 88 | |
| BC066527.1 | - | 6 - 7 | 600 | 88 | ||
| BC091971.1 | - | 6 - 7 | 622 | 88 | ||
More Information
Set Membership: |
|

bzw1b
Genomic Map