Assay Details
| Gene Symbol: bzw1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213092.1 | NP_998257.1 | 9 - 10 | 1027 | 129 | |
| XM_005167390.4 | XP_005167447.1 | 9 - 10 | 1117 | 129 | ||
| GenBank mRNA | BC066527.1 | - | 9 - 10 | 1027 | 129 | |
| BC091971.1 | - | 9 - 10 | 1049 | 129 | ||
More Information
Set Membership: |
|

bzw1b
Genomic Map