Assay Details
| Gene Symbol: slc20a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213179.1 | NP_998344.1 | 2 - 3 | 698 | 61 | |
| XM_005167150.4 | XP_005167207.1 | 2 - 3 | 778 | 61 | ||
| GenBank mRNA | BC068354.1 | - | 2 - 3 | 698 | 61 | |
More Information
Set Membership: |
|

slc20a1a
Genomic Map