Assay Details
| Gene Symbol: prps1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213327.2 | NP_998492.1 | 2 - 3 | 481 | 102 | |
| NM_213533.3 | NP_998698.1 | 2 - 3 | 481 | 102 | ||
| XM_009301280.3 | XP_009299555.1 | 2 - 3 | 513 | 102 | ||
| GenBank mRNA | BC045301.1 | - | 2 - 3 | 453 | 102 | |
| BC066485.1 | - | 2 - 3 | 465 | 102 | ||
| CT644358.2 | - | 1 - 2 | 163 | 102 | ||
More Information
Set Membership: |
|

prps1a
Genomic Map