Assay Details
| Gene Symbol: prps1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001359894.1 | NP_001346823.1 | 3 - 4 | 582 | 87 | |
| NM_213327.2 | NP_998492.1 | 3 - 4 | 576 | 87 | ||
| NM_213533.3 | NP_998698.1 | 3 - 4 | 576 | 87 | ||
| XM_009301279.3 | XP_009299554.1 | 3 - 4 | 616 | 87 | ||
| XM_009301280.3 | XP_009299555.1 | 3 - 4 | 608 | 87 | ||
| XM_021475990.1 | XP_021331665.1 | 2 - 3 | 121 | 87 | ||
| GenBank mRNA | BC045301.1 | - | 3 - 4 | 548 | 87 | |
| BC066485.1 | - | 3 - 4 | 560 | 87 | ||
| BC098599.1 | - | 3 - 4 | 581 | 87 | ||
| CT644358.2 | - | 2 - 3 | 258 | 87 | ||
More Information
Set Membership: |
|

prps1a
Genomic Map