Assay Details
| Gene Symbol: cdc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213386.1 | NP_998551.1 | 10 - 11 | 934 | 67 | |
| GenBank mRNA | BC050511.1 | - | 10 - 11 | 967 | 67 | |
| BC154772.1 | - | 10 - 11 | 947 | 67 | ||
More Information
Set Membership: |
|

cdc45
Genomic Map