Assay Details
| Gene Symbol: ndufa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002153.3 | NP_001002153.2 | 2 - 3 | 381 | 94 | |
| NM_001167855.2 | NP_001161327.1 | 3 - 4 | 292 | 94 | ||
| GenBank mRNA | BC071358.1 | - | 2 - 3 | 281 | 94 | |
| EH451895.1 | - | 3 - 4 | 254 | 94 | ||
More Information
Set Membership: |
|

ndufa1
Genomic Map