Assay Details
| Gene Symbol: mpp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002223.1 | NP_001002223.1 | 11 - 12 | 1710 | 136 | |
| XM_005156066.4 | XP_005156123.1 | 10 - 11 | 1649 | 136 | ||
| XM_009306493.3 | XP_009304768.1 | 10 - 11 | 1634 | 136 | ||
| XM_009306494.2 | XP_009304769.1 | 9 - 10 | 1580 | 136 | ||
| GenBank mRNA | BC074066.1 | - | 11 - 12 | 1710 | 136 | |
| BC165715.1 | - | 10 - 11 | 1377 | 136 | ||
More Information
Set Membership: |
|

mpp2b
Genomic Map