Assay Details
| Gene Symbol: rab15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002318.1 | NP_001002318.1 | 3 - 4 | 408 | 114 | |
| GenBank mRNA | BC075754.1 | - | 3 - 4 | 408 | 114 | |
More Information
Set Membership: |
|

rab15
Genomic Map