Assay Details
| Gene Symbol: rgs20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002431.2 | NP_001002431.1 | 3 - 4 | 685 | 79 | |
| XM_005163426.4 | XP_005163483.1 | 3 - 4 | 725 | 79 | ||
| XM_005163427.4 | XP_005163484.1 | 3 - 4 | 713 | 79 | ||
| XM_005163428.4 | XP_005163485.1 | 3 - 4 | 365 | 79 | ||
| XM_009298568.1 | XP_009296843.1 | 3 - 4 | 325 | 79 | ||
| GenBank mRNA | BC076135.1 | - | 3 - 4 | 686 | 79 | |
More Information
Set Membership: |
|

rgs20
Genomic Map