Assay Details
| Gene Symbol: naxe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002618.1 | NP_001002618.1 | 4 - 5 | 572 | 73 | |
| GenBank mRNA | BC075969.1 | - | 4 - 5 | 572 | 73 | |
More Information
Set Membership: |
|

naxe
Genomic Map