Assay Details
Gene Symbol: prkab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002632.1 | NP_001002632.1 | 3 - 4 | 421 | 107 | |
XM_005155628.4 | XP_005155685.1 | 3 - 4 | 488 | 107 | ||
GenBank mRNA | BC075947.1 | - | 3 - 4 | 421 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
