Assay Details
Gene Symbol: prkab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002632.1 | NP_001002632.1 | 5 - 6 | 632 | 80 | |
XM_005155628.4 | XP_005155685.1 | 5 - 6 | 699 | 80 | ||
GenBank mRNA | BC075947.1 | - | 5 - 6 | 632 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
